L(+)-Tartaric acid

Supplier: RPI
Danger

T78350-1000.0 T78350-500.0 T78350-5000.0
77510-488EA 66.25 USD
77510-488 77510-490 77510-492
L(+)-Tartaric acid
L(+)-Tartaric acid

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₄H₆O₆
Melting Pt: 170...172 °C
Flash Pt: 150 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0

Order Now

Specification Test Results

Appearance Colorless or translucent or a white powder
Arsenic (As) ≤2 ppm
Assay (Dried Basis) 99.7 - 100.5%
Heavy Metals (as Pb) ≤2 ppm
Loss on Drying ≤0.5%
Oxalate Test Pass Test
Residue on Ignition ≤0.05%
Specific Rotation +12.0 - +13.0°
Sulfate Test Pass Test
BSE/TSE Free BSE/TSE Free
Country of Origin CN


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR