Tris-Glycine Buffer 10X Powder
Supplier: RPI
T29040-1.0
77510-458EA 27.78 USD
77510-458
Tris-Glycine Buffer 10X Powder
Buffers Biological Buffers
Dry homogeneous blend of molecular biology grade TRIS base and Glycine. Add 174.4 g to make 1 L of 10X concentrate.
Caution: Research or further manufacturing use only, not for food or drug use.