Nutrient Agar, Powder
Supplier: RPI
For cultivating a variety of non-fastidious organisms.
- Ingredients:
- Beef extract: 3 g/L
- Pancreatic digest of gelatin: 5 g/L
- Agar: 15 g/L
Dissolve 23 g in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121 °C for 15 minutes. Mix medium with gentle agitation to avoid bubbles prior to pouring plates. Store at room temperature.
Caution: Research or further manufacturing use only, not for food or drug use.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...