MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)

Supplier: RPI

M81950-1000.0 M81950-2000.0 M81950-250.0 M81950-500.0
77510-334EA 860.56 USD
77510-334 77510-336 77510-338 77510-340
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₇H₁₄NNaO₅S
Storage Temperature: Room Temperature
MDL Number: MFCD00070007
CAS Number: 79803-73-9

Order Now

Specification Test Results

Appearance White crystalline powder
Solubility (10% in H₂O) Clear and colorless
pH 8.9 - 9.7
Absorbance (260 nm) (5% aqueous) ≤0.04
Absorbance (280 nm) (5% aqueous) ≤0.02
Loss on Drying ≤1.0%
Assay (by titration) ≥99.0%
Residual Solvent (Ethanol Class III) Complies with ICH Q3C (R5)
Melamine Free Melamine Free
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR