LB Broth, Miller's, Liquid (Luria-Bertani Broth)
Supplier: RPI
LB Broth Miller's Liquid is a fundamental growth medium essential for bacterial cultivation. Specifically formulated, it provides an optimal nutrient blend to support robust bacterial growth.
- Tryptone: 10.0 g/L
- Yeast extract: 5.0 g/L
- Sodium chloride: 10.0 g/L
Widely preferred for its reliability and versatility, LB Broth Miller's Liquid is compatible with a diverse array of bacterial strains, making it indispensable for microbiology, molecular biology, and biotechnology applications. Its balanced formulation fosters rapid bacterial proliferation, facilitating genetic manipulation, protein expression studies, and other research endeavors. Researchers rely on LB Broth Miller's Liquid as a foundational tool to explore microbial physiology, genetics, and the mechanisms underlying various biological processes.
Caution: Research or further manufacturing use only, not for food or drug use.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...