D-(-)-Luciferin

Supplier: RPI

L37000-0.5
77510-302EA 569.79 USD
77510-302
D-(-)-Luciferin
D(-)-Luciferin

Used with firefly luciferase in bioluminescence for the determination of ATP.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₁H₈N₂O₃S₂
Melting Pt: 201 °C
Storage Temperature: Freezer
MDL Number: MFCD00042929
CAS Number: 2591-17-5
EINECS: 219-981-3

Order Now

Specification Test Results

Appearance White to yellow powder
Biological Activity (Basic, ABS ratio of 385/285 nm) ≥2.5
Biological Activity (Neutral, ABS ratio of 327/268 nm) ≥2.3
Identification (FTIR) Conforms to structure
Purity (HPLC) ≥98%
Chiral Purity ≥97.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR