Guanidinium thiocyanate
Supplier: RPI
|
Danger
|
Synonyms:
Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
G54000-100.0
G54000-1000.0
G54000-500.0
G54000-5000.0
G54000-250.0
76346-200EA
63.36
USD
76346-200
76346-202
76346-204
76346-206
77510-268
Guanidinium thiocyanate
Guanidinium thiocyanate
Protein denaturant used for purifying proteins and nucleic acids.
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
CH₅N₃·CHNS Storage Temperature: Room Temperature |
MDL Number:
MFCD00013027 CAS Number: 593-84-0 EINECS: 209-812-1 |
Specification Test Results
Appearance | White to slightly off white crystalline powder |
Assay (Argentometric Titration) | ≥ 99.0 % |
Assay (Ion Chromatography) | ≥ 99.5 % |
Chloride Content | ≤ 1.0 % |
Cobalt (Co) | ≤ 5 ppm |
FTIR | Conforms to reference |
Iron | ≤ 5 ppm |
pH | 5.0 - 7.0 |
DNase/ RNase | None Detected |
Melting Point | 118 - 121 oC |
Solubility | Pass |
Sulfate Ash | ≤ 0.5 % |
Moisture | ≤ 0.5 % |
BSE/TSE Free | BSE/TSE Free |
Grade | Ultra Pure |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...