Formamide

Supplier: RPI
Danger

Synonyms: Carbamaldehyde

F11900-0.1 F11900-0.5 F11900-1000.0
76345-988EA 24.83 USD
76345-988 76345-990 77510-258
Formamide
Formamide

For nucleic acid hybridization and sequencing.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: HCONH₂
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0

Order Now

Specification Test Results

Appearance Colorless liquid
Assay ≥ 99.5 %
Freezing Point 1.0 - 3.0 °C
Copper ≤ 0.0001 %
Iron ≤ 0.0005 %
Lead (Pb) ≤ 0.0001 %
Zinc ≤ 0.0001 %
Country of Origin DE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR