kappa-Carrageenan

Supplier: RPI

C41070-100.0 C41070-1000.0 C41070-250.0 C41070-5000.0
76345-492EA 39.4 USD
76345-492 76345-494 76345-496 77510-220
kappa-Carrageenan
kappa-Carrageenan

Kappa carrageenan binds water to form strong, rigid gels in the presence of potassium salts. As the level of potassium is increased, the gel structure becomes tightly aggregated and may result in syneresis (moisture on the gel surface).


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₂H₁₇O₁₃S
Storage Temperature: Room Temperature
MDL Number: MFCD00151514
CAS Number: 11114-20-8

Order Now

Specification Test Results

Appearance White to Light Cream
Yeast and Mold ≤100 cfu/g
Loss on Drying (70 °C, 18 hr) ≤12.5%
Aerobic Plate Count ≤200 CFU/g
Salmonella Negative (In 25 g)
E.Coli Negative (In 12.5 g)
pH (1% Solution) 8.0 - 11.0
Breaking Strength (0.7%) 300 - 700 g TA
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR