Citric acid in ACS Reagent grade water
Supplier: RPI
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
C34525-500.0
C34525-3000.0
C34525-5000.0
C34525-1000.0
76345-416EA
39.4
USD
76345-416
77510-212
77510-214
77510-210
Citric acid in ACS Reagent grade water
Citric acid
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 310 Melting Pt: 153...159 °C Storage Temperature: Room Temperature |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 Merck Index: 13,02350 |
Specification Test Results
Appearance | White Crystals |
IR spectrum | Conforms to structure |
Purity (Titration by NaOH) | ≥99.50% |
Chloride Content | ≤0.001% |
Insoluble Matter | ≤0.005% |
Iron | ≤3 ppm |
Lead | ≤2 ppm |
Oxalate | Pass |
Phosphate | ≤0.001% |
Residue on Ignition (Ash) | ≤0.02% |
Subs Carbonizable by Hot H2SO4 | Pass |
Sulfur Compounds | ≤0.002% |
Grade | ACS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...