Citric acid, anhydrous
Supplier: RPI
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
C34500-1000.0
C34500-500.0
C34500-5000.0
C34500-10000.0
C34500-25000.0
76345-410EA
65.99
USD
76345-410
76345-412
76345-414
77510-206
77510-208
Citric acid, anhydrous
Citric acid
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 310 Melting Pt: 153...159 °C Storage Temperature: Room Temperature |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 Merck Index: 13,02350 |
Specification Test Results
Appearance | Fine white crystalline powder |
Assay (Titration) | ≥99.5 % |
Carbonizable Substances (H2SO4) | Pass Test |
Clarity and Color | Pass Test |
Identification | Pass Test |
Lead | ≤0.5 mg/kg |
Limit of Oxalic Acid | ≤0.036 % |
Oxalate Test | Pass Test |
Residual Solvents | Meet requirements |
Residue on Ignition | ≤0.05 % |
Sulfate | ≤0.015 % |
Water | ≤0.5 % |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...