Sealed Air® Bubble Wrap® Cushioning, Anti-Static Multi-Purpose Grade, Pink Tinted
Supplier: Associated Bag
505-23-22
77510-130RL 305.9 USD
77510-130
Sealed Air® Bubble Wrap® Cushioning, Anti-Static Multi-Purpose Grade, Pink Tinted
Seals General Purpose Sealing Films
Anti-static bubble roll.
  • Made with an air-retaining nylon barrier surrounded by layers of specially treated anti-static polyethylene
Sealed Air® Bubble Wrap® multi-purpose grade cushioning (³/₁₆") is great for general packaging and short shipping cycles. Air retention barrier is co-extruded with two layers of polyethylene, 30% fuller than other bubble roll. Perforated every 12".