pDNA Resuspension Buffer, pH 8.0
Supplier: TEKNOVA INC
21216
77509-556EA 76.49 USD
77509-556
pDNA Resuspension Buffer, pH 8.0
Buffers Biological Buffers
pDNA Resuspension buffer composition: 50 mM Tris/Tris-HCl, 10 mM EDTA.