L-Shaped Spreader
Supplier: NEST SCIENTIFIC
711001
77508-584CS 268.64 USD
77508-584
L-Shaped Spreader
Spreaders Cell Spreaders
Using a grinding tool with high precision in combination with an ergonomic design, the efficient sterile L-Spreader specialized for Asian scientific researchers is developed and manufactured to effectively solve problems such as contamination or uneven application of spread plate in bio-labs, which helps to make domestic biological testing
practitioners more efficient and simplify their tedious work during biological experiments.
  • Sterile L-Spreader, using medical PS
  • EO Sterilized
  • Smooth and flat surface that solves problems such as contamination or uneven application of spread plate in bio-labs
  • Length of handle: 144 mm, diagram of spreading: 33 mm
  • Ideal for spreading cells or bacteria evenly in the culture dish
Suitable for 3.5, 6, 9 and 10 cm petri dishes and more.