Centrifuge Tubes
Supplier: NEST SCIENTIFIC
601151
77508-218CS 120.02 USD
77508-218
Centrifuge Tubes
Tubes Centrifuge Tubes
The 15 ml Centrifuge Tube by NEST Scientific is a versatile lab essential. Sold in bulk with 50 tubes per pack and 500 per case, these standard tubes are non-sterile with caps separated for convenience. Ideal for various laboratory applications, these tubes are compatible with most standard centrifuges, allowing for efficient sample processing.
  • Made with high-quality materials
  • These tubes offer reliable performance and durability
Whether used for sample storage, centrifugation, or mixing applications, these 15 ml centrifuge tubes are a practical choice for any laboratory setting. Stock up on these tubes to meet your daily lab needs with ease.