VWR® Lab Pen black ink
Supplier: VWR International
SMP-BK/109
77507-062PK 89.32 USD
77507-062
VWR® Lab Pen black ink
Pens Permanent Markers
VWR®, part of Avantor has industry leading Anatomical Pathology consumables available your lab.
  • Designed exclusively for lab techs, this original formulation does not dry out, and withstands exposure to common processing chemicals.
  • Excellent for both slides and embedding cassettes.