Xylene (mixture of isomers), Lab-Grade, VWR®

Supplier: VWR International
Danger

Synonyms: Dimethylbenzene

77507-038
77507-038CS 298.41 USD
77507-038
Xylene (mixture of isomers), Lab-Grade, VWR®
Xylene (mixture of isomers)

VWR®, part of Avantor has industry leading Anatomical Pathology reagents available for your lab.


  • ISO 13485 Certified
  • F-style gallons for easier storage and fewer spills
  • Transparency and availability to help you with reliability


Certifications: ISO 13485 Certified

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –34 °C
Flash Pt: 25 °C (77 °F)
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR