2-Propanol, VWR®

Supplier: VWR International
Danger

77507-026
77507-026CS 175.14 USD
77507-026
2-Propanol, VWR®
2-Propanol

VWR®, part of Avantor has industry leading Anatomical Pathology reagents available for your lab.


  • ISO 13485 Certified
  • F-style gallons for easier storage and fewer spills
  • Transparency and availability to help you with reliability


Certifications: ISO 13485 Certified.


Delivery information: Supplied with screw cap and pouring ring.


Packaging: The bottles are packaged in shrinked-wrapped modules.

Formula: (CH₃)₂CHOH
Boiling Pt: 82…82.3 °C (179.6…180.14 °F)
Flash Pt: 12 - 13 °C (53.6 - 55.4 °F)
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR