Indicator tape for formaldehyde sterilization processes.
- Size: 50 m × 18 mm
- Single-use chemical indicators that consist of a roll of self-adhesive plastic tape printed with indicator ink
- Designed to monitor Plasma or Vaporized Hydrogen Peroxide sterilization processes, ensuring an adequate exposure to the sterilizing agent during the sterilization process and to distinguish between processed and unprocessed items
- The adhesive component of the tape allows adhesion to different types of packaging and wraps, such as cloth, paper and plastic being easy to remove and leaves no residues
- Innovative highly sensitive purple ink was developed to turn to green when the indicator is exposed to Plasma or Vaporized Hydrogen Peroxide
- 100% free of toxic heavy metals
For monitoring Formaldehyde sterilization processes. External use. Conditions: 15 minutes, 70 °C, 1.0 mol/l FORM.
Storage conditions:
Temperature: 10 to 30 °C, RH: 30-80%, keep out of direct light.
Certifications: ISO 13485; ISO 11140-1.
Packaging: 1 roll per package.