Single Hopper Thermal Cassette Printer, PiSmart
Supplier: StatLab
PISMART1HOPPER-PSPKIT
77506-764EA 35231.78 USD
77506-764
Single Hopper Thermal Cassette Printer, PiSmart
Printers Cassette and Slide Printers
Designed for on-demand printing, hoppers on the the PiSmart single can easily swap out, or the hoppers can be bypassed to manually drop in a different type of cassette. High-quality thermal transfer technology delivers a dark, high-contrast print.
  • Industry smallest footprint
  • A fast, 3 - 5 second print time
  • Extremely quiet!
  • Easy-to-learn interface
  • Dark, high-contrast barcode printing
  • Allows up to 100 characters on each cassette
  • Built in scanner
  • LIS integration delivers a smart solution
  • Four years PiSmart promise warranty
Control all of your PiSmartMore Product Information