Syringe for Automatic Liquid Handling, INNOVADYNE* NANODROP**
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2623917
77506-330EA 181.51 USD
77506-330
Syringe for Automatic Liquid Handling, INNOVADYNE* NANODROP**
Syringes Syringes for Chromatography
The glass barrel of the syringes are made up of borosilicate 3.3.
  • 60 mm stroke
  • Series: H
  • Plunger tip: PTFE