Syringes for Thin Layer Chromatography
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2606702
77506-154EA 152.16 USD
77506-154
Syringes for Thin Layer Chromatography
Syringes Syringes for Chromatography
This syringe is specially designed for thin layer chromatography.
  • Glass barrel is made of Borosilicate 3.3
  • Precision: <±1% of the volume
The needle is developed for the needs of the e Linomat III and IV.