Syringe for Automatic Liquid Handling, AD
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2605813
77506-152EA 80.35 USD
77506-152
Syringe for Automatic Liquid Handling, AD
Syringes General-Purpose Syringes
Syringes for automatic liquid handling made up of borosilicate 3.3.
  • 60 mm stroke
Precision: < ±1% of the volume
Front fitting: M8×0.75