Syringes for HPLC Autosamplers
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2601702
77506-150EA 61.77 USD
77506-150
Syringes for HPLC Autosamplers
Syringes Syringes for Chromatography
  • HPLC valves
  • Autosamplers
  • Dosage of aggressive fluids
  • Glass barrel: Borosilicate 3.3
  • Plunger tip: PTFE
Plunger: Stainless steel with PTFE-seal
Glass barrel: Borosilicate 3.3