3-Formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10563-25G
77497-578EA 64.38 USD
77497-578
3-Formylphenylboronic acid ≥97%
3-Formylphenylboronic acid

Caution: for Research Use Only

Formula: C₇H₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD00161356
CAS Number: 87199-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR