4-Hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

10494-25G
77497-538EA 64.38 USD
77497-538
4-Hydroxyphenylboronic acid ≥97%
4-Hydroxyphenylboronic acid

Caution: for Research Use Only

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR