4-(Trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10447-25G
77497-514EA 64.38 USD
77497-514
4-(Trifluoromethyl)phenylboronic acid ≥97%
4-(Trifluoromethyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₇H₆BF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR