2-Methoxy-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

10274-25G
77497-452EA 64.38 USD
77497-452
2-Methoxy-5-pyridineboronic acid ≥97%
2-Methoxy-5-pyridineboronic acid

Caution: for Research Use Only

Formula: C₆H₈BNO₃
Storage Temperature: Ambient
MDL Number: MFCD02093044
CAS Number: 163105-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR