3-Chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10231-25G
77497-406EA 64.38 USD
77497-406
3-Chlorophenylboronic acid ≥97%
3-Chlorophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BClO₂
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR