2-Chloro-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

10203-25G
77497-392EA 64.38 USD
77497-392
2-Chloro-3-pyridineboronic acid ≥97%
2-Chloro-3-pyridineboronic acid

Caution: for Research Use Only

Formula: C₅H₅BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD03094997
CAS Number: 381248-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR