4-Carboxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

10183-25G
77497-384EA 64.38 USD
77497-384
4-Carboxyphenylboronic acid pinacol ester ≥97%
4-Carboxyphenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₃H₁₇BO₄
Storage Temperature: Ambient
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR