2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%
Supplier: AOBChem USA
319801-1G
319801-500MG
319801-5G
319801-25G
77469-094EA
415.09
USD
77469-094
77481-770
77492-168
77502-100
2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone
Caution: For research use only
Formula:
C₁₇H₁₅ClF₂O₂ |
CAS Number:
3002454-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...