1-(3-(tert-Butyl)phenyl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

99380-1G 99380-500MG 99380-5G 99380-25G
77473-968EA 415.09 USD
77473-968 77486-578 77497-204 77506-088
1-(3-(tert-Butyl)phenyl)-4,4-dimethylcyclohexanol ≥97%
1-(3-(tert-Butyl)phenyl)-4,4-dimethylcyclohexanol

Caution: For research use only

Formula: C₁₈H₂₈O
Storage Temperature: Ambient
MDL Number: MFCD34775993
CAS Number: 2918917-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR