2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

319801-1G 319801-500MG 319801-5G 319801-25G
77469-094EA 415.09 USD
77469-094 77481-770 77492-168 77502-100
2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-methyl-4-((4-methylbenzyl)oxy)phenyl)ethanone

Caution: For research use only

Formula: C₁₇H₁₅ClF₂O₂
CAS Number: 3002454-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR