VWR® Integrated Cleanroom Hood and Mask, Made in USA
Supplier: VWR International
449
77474-002CS 1234.82 USD
77474-002
VWR® Integrated Cleanroom Hood and Mask, Made in USA
Disposable Garments
The VWR® integrated hood and mask is an ideal PPE solution for use in cleanroom and sterile environments.
  • Innovative design
  • Built-in tie-back
  • Unique crown design for ulitmate comfort
  • Designed with eyewear slots to hold glasses or goggles in place
  • Made in USA
This innovative garment combines a hood with a built-in tie-back mask, ensuring comprehensive protection for users. The unique crown design provides the perfect user fit for all sizes. Featuring convenient eyewear arm slots, it securely holds glasses or goggles in place, enhancing safety and comfort. These eyeglass slots also keep the hood securely placed against the face, allowing wearers to stay shielded and focused on their work.
Packaging: Packaged 25 per bag, 4 bags per case (100 units per case).