Non-Contact Dispensers, I.DOT
Supplier: DISPENDIX CELLINK LLC
D16110024973 D16110026000 D16110021796 D16110021802 D16110021808 D16110021814 D16110022091 D16110024910
77463-752PK 3950.03 USD
77463-752 77463-754 77463-732 77463-734 77463-736 77463-738 77463-742 77463-748
Non-Contact Dispensers, I.DOT
Automated Liquid Handling Systems
An accessible research appliance enabling the dispensing of nanoliter and microliter scale volumes to 96-, 384-, and 1536-well plates.
  • Use a fraction of the pipette tips required by traditional automation
  • Dispense reagents with a 1 μl dead volume and run more samples for the same reagent cost
  • Dispense 4 nl to 30 μl into any SBS format destination labware with a high degree of precision
  • Dispense with insight into the actual volume dispensed every time with integrated volume verification
  • Software for dispensing a constant volume, dilution, normalization, and
More Product Information