GemCell™ Plus Xeno-Free Human Serum, World Grade
Supplier: GeminiBio
100-916-100
77463-600EA 585.34 USD
77463-600
GemCell™ Plus Xeno-Free Human Serum, World Grade
Sera (Serum)
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.
  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0.1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Meet US and EU regulatory requirements