Human Serum AB (Off The Clot), Gamma Irradiated
Supplier: GeminiBio
100-318G-100
77463-596EA 789.32 USD
77463-596
Human Serum AB (Off The Clot), Gamma Irradiated
Sera (Serum)
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
  • Supports clinical and commercial applications
  • Aseptic assurance system
  • Gamma irradiated