BenchCel Workstation
Supplier: AGILENT TECHNOLOGIES, INC
G5590GA
77463-590EA 89307.18 USD
77463-590
BenchCel Workstation
Automated Liquid Handling Systems
The Agilent BenchCel workstation is an automated microplate handler capable of transferring multiple plates at once from a microplate stacker to utilities enabling barcode and other plate labeling, plate centrifugation, and plate sealing. The compact workstation can be used in any lab setting.
  • Two, four, or six labware rack configurations available with the BenchCel microplate handler enhance scalability
  • Platform can be configured with the Agilent PlateLoc thermal microplate sealer, Microplate barcode labeler, or microplate centrifuge
  • Enabled for microplate workflows requiring automated plate sealing, barcode labeling, and plate centrifugation
  • Eight-second transfer time from stack to instrument
  • Adjustable plate capacity handles up to 60 standard SBS microplates per rack
Hands-on time is reduced to a minimum through the automation of routine yet laborious manual microplate management tasks, increasing your lab’s productivity. Save further on setup time with intuitive VWorks software, allowing you to perform microplate stacking and sealing management tasks with confidence and efficiency.