PIG® Aggressive Chemical Absorbent Packaging Pouches
Supplier: New Pig
PKG3016
77463-560BX 71.1 USD
77463-560
PIG® Aggressive Chemical Absorbent Packaging Pouches
Mailers and Shippers
Protect against aggressive-chemical leaks during shipping, as well as against scratching and light shifting, with these absorbent pouches.
  • Heavyweight absorbent sheet is perfectly sewn into a pouch to soak up aggressive chemicals
  • Packaging pouch also cushions against light shifting
  • Material is soft and non-abrasive to protect from scratching
  • Soaks up most aggressive chemicals
  • Durable multi-layer construction holds together, even when saturated
  • Packaging absorbent that is still dry upon shipment arrival is reusable for sustainability
Absorbency up to 5.05 gal. per box, absorbency per pouch up to 25.86 oz.