Fixing Plug Set for Upright Tubes
Supplier: VWR International
77463-236
77463-236PK 35.75 USD
77463-236
Fixing Plug Set for Upright Tubes
Tool Sets
Fixing plug set includes two plugs. These plugs are needed when mounting horizontal profiles to the upright tubes, two plug per upright tube.
  • Colored bioplastic with fiber reinforcement
Ordering information: Installation is required. Assembly videos available.