Cy5 aldehyde
Supplier: AAT BIOQUEST INC
9012
77463-120EA 216.1 USD
77463-120
Cy5 aldehyde
Cy5 aldehyde
Cy5 aldehyde is a reactive red fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only