iFluor 700 SSL-NHS ester, ReadiCleave™
Supplier: AAT Bioquest
7053
77463-048EA 374.15 USD
77463-048
iFluor 700 SSL-NHS ester, ReadiCleave™
iFluor 700 SSL-NHS ester
ReadiCleave™ iFluor® 700 succinimidyl ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the iFluor® 700 fluorophore from the target molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Caution: For Research Use Only