Cy5 Dye qPCR Calibration Solution *10000X*
Supplier: AAT BIOQUEST INC
67033
77463-012EA 318.85 USD
77463-012
Cy5 Dye qPCR Calibration Solution *10000X*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.