FAM Dye qPCR Calibration Solution *10000X*
Supplier: AAT BIOQUEST INC
67032
77463-010EA 318.85 USD
77463-010
FAM Dye qPCR Calibration Solution *10000X*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
FAM dye qPCR calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Calibration and verification should be run every six months or as needed.
Caution: For research use only.