NED Dye qPCR Calibration Solution *10000X*
Supplier: AAT BIOQUEST INC
67031
77463-008EA 318.85 USD
77463-008
NED Dye qPCR Calibration Solution *10000X*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
NED dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.