TAMRA Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Supplier: AAT BIOQUEST INC
67000
77462-986EA 101.72 USD
77462-986
TAMRA Dye qPCR Calibration Plate *Optimized for ABI7500 Fast 96-Well*
Microplates PCR Plates
The TAMRA dye qPCR calibration plate can be used to calibrate and maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than three times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimize light exposure.