Cy5 phosphoramidite
Supplier: AAT BIOQUEST INC
6071
77462-966EA 215.09 USD
77462-966
Cy5 phosphoramidite
Cy5 phosphoramidite
Two cyanine derivatives, Cy3 and Cy5 phosphoramidites, are the most popular fluorescent labeling reagents for preparing fluorescence-labeled oligonucleotides.
  • Stable for 6 months
Quality tested in-house
Caution: For Research Use Only