Transfectamine™ 5000 Transfection Reagent
Supplier: AAT BIOQUEST INC
60019
77462-936EA 41.85 USD
77462-936
Transfectamine™ 5000 Transfection Reagent
Transfection Reagents
Transfectamine™ 5000 Transfection Reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells.
  • Consistently high transfection efficiency
  • Higher viability of transfected cells
  • Does not require special medium
Transfectamine™ 5000 is easier to use compare to most other transfection reagents and does not require special medium.
Caution: For research use only.