ReadiLink™ Rapid XFD555 Antibody Labelling Kit
Supplier: AAT BIOQUEST INC
5733
77462-928EA 523.82 USD
77462-928
ReadiLink™ Rapid XFD555 Antibody Labelling Kit
Protein Linking and Labeling Systems
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped at RT.
Caution: Research use only.